Gene Transcripts
- Gene Symbol
- CCNDBP1
- Entrez Gene ID
- 23582
- Gene Name
- cyclin D1 binding protein 1
- Gene Aliases
- DIP1, GCIP, HHM
- Chromosome Location
- Chr. 15: 43185118 - 43197177 on Build GRCh38
- UniGene ID
- Hs.36794
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012142.4 |
NP_036274.3
|
8, 9 |
1089 |
|
NR_027513.2 |
|
6, 7 |
912 |
|
NR_027514.2 |
|
7, 8 |
992 |
|
NR_045998.1 |
|
7, 8 |
1009 |
|
NR_045999.1 |
|
5, 6 |
852 |
|
XM_006720448.1 |
XP_006720511.1
|
6, 7 |
1011 |
|
GenBank mRNA |
AF082569.1 |
- |
8, 9 |
948 |
|
AF087852.1 |
- |
8, 9 |
934 |
|
AF113535.1 |
- |
6, 7 |
898 |
|
AF132034.1 |
- |
8, 9 |
904 |
|
AF246144.1 |
- |
8, 9 |
906 |
|
AK075146.1 |
- |
8, 9 |
918 |
|
AK075296.1 |
- |
6, 7 |
903 |
|
AK124193.1 |
- |
3, 4 |
1493 |
|
AK128849.1 |
- |
4, 5 |
2133 |
|
AK225143.1 |
- |
8, 9 |
904 |
|
AK225645.1 |
- |
8, 9 |
918 |
|
AK290665.1 |
- |
8, 9 |
918 |
|
AK290707.1 |
- |
7, 8 |
871 |
|
AK295017.1 |
- |
7, 8 |
821 |
|
BC009689.1 |
- |
8, 9 |
932 |
3896421 |
BQ439439.1 |
- |
5, 6 |
666 |
|
CR450331.1 |
- |
8, 9 |
858 |
|
View Full Product Details