Gene Transcripts
- Gene Symbol
- TMEM9
- Entrez Gene ID
- 252839
- Gene Name
- transmembrane protein 9
- Gene Aliases
- DERM4, TMEM9A
- Chromosome Location
- Chr. 1: 201134771 - 201171625 on Build GRCh38
- UniGene ID
- Hs.181444
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001288564.1 |
NP_001275493.1
|
2, 3 |
195 |
|
NM_001288565.1 |
NP_001275494.1
|
1, 2 |
698 |
|
NM_001288566.1 |
NP_001275495.1
|
2, 3 |
296 |
|
NM_001288567.1 |
NP_001275496.1
|
2, 3 |
510 |
|
NM_001288568.1 |
NP_001275497.1
|
2, 3 |
348 |
|
NM_001288569.1 |
NP_001275498.1
|
2, 3 |
209 |
|
NM_016456.4 |
NP_057540.1
|
2, 3 |
213 |
|
NR_109920.1 |
|
2, 3 |
213 |
|
NR_109921.1 |
|
1, 2 |
698 |
|
GenBank mRNA |
AB013909.1 |
- |
1, 2 |
66 |
|
AF151020.1 |
- |
3, 4 |
185 |
|
AK075335.1 |
- |
2, 3 |
123 |
|
AK091726.1 |
- |
1, 2 |
583 |
|
AK296307.1 |
- |
2, 3 |
130 |
|
AL136600.1 |
- |
2, 3 |
146 |
|
AY138587.1 |
- |
2, 3 |
124 |
|
AY359012.1 |
- |
2, 3 |
187 |
|
BC001106.2 |
- |
2, 3 |
137 |
3506442 |
BE782946.1 |
- |
2, 3 |
175 |
|
BM928361.1 |
- |
2, 3 |
135 |
|
BX337142.2 |
- |
1, 2 |
582 |
|
CR457162.1 |
- |
1, 2 |
66 |
|
CR533467.1 |
- |
1, 2 |
66 |
|
CX751563.1 |
- |
1, 2 |
580 |
|
DA141558.1 |
- |
2, 3 |
195 |
|
DA364005.1 |
- |
2, 3 |
213 |
|
DB031447.1 |
- |
2, 3 |
189 |
|
View Full Product Details