Gene Transcripts
- Gene Symbol
- GOLGA3
- Entrez Gene ID
- 2802
- Gene Name
- golgin A3
- Gene Aliases
- GCP170, MEA-2
- Chromosome Location
- Chr. 12: 132768909 - 132829131 on Build GRCh38
- UniGene ID
- Hs.507333
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001172557.1 |
NP_001166028.1
|
6, 7 |
1505 |
|
NM_005895.3 |
NP_005886.2
|
6, 7 |
1843 |
|
XM_005266164.4 |
XP_005266221.1
|
6, 7 |
1450 |
|
XM_005266167.4 |
XP_005266224.1
|
3, 4 |
875 |
|
XM_006719736.3 |
XP_006719799.1
|
6, 7 |
1570 |
|
XM_006719737.3 |
XP_006719800.1
|
6, 7 |
1580 |
|
XM_011534793.2 |
XP_011533095.1
|
5, 6 |
1297 |
|
GenBank mRNA |
AB027133.1 |
- |
6, 7 |
1565 |
|
AF485338.1 |
- |
5, 6 |
1287 |
|
BC060826.1 |
- |
6, 7 |
1529 |
|
BC142658.1 |
- |
6, 7 |
1505 |
40148073 |
BC146675.1 |
- |
6, 7 |
1505 |
40148852 |
D63997.1 |
- |
6, 7 |
1553 |
|
View Full Product Details