Gene Transcripts
- Gene Symbol
- EPHA6
- Entrez Gene ID
- 285220
- Gene Name
- EPH receptor A6
- Gene Aliases
- EHK-2, EHK2, EK12, EPA6, HEK12, PRO57066
- Chromosome Location
- Chr. 3: 96814581 - 97761532 on Build GRCh38
- UniGene ID
- Hs.272208
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001080448.2 |
NP_001073917.2
|
8, 9 |
2049 |
|
NM_001278300.1 |
NP_001265229.1
|
5, 6 |
461 |
|
NM_173655.3 |
NP_775926.1
|
6, 7 |
577 |
|
XM_006713592.3 |
XP_006713655.1
|
9, 10 |
2097 |
|
XM_017006210.1 |
XP_016861699.1
|
9, 10 |
2097 |
|
XM_017006211.1 |
XP_016861700.1
|
10, 11 |
2122 |
|
XM_017006212.1 |
XP_016861701.1
|
9, 10 |
1791 |
|
XM_017006213.1 |
XP_016861702.1
|
9, 10 |
2149 |
|
XM_017006214.1 |
XP_016861703.1
|
8, 9 |
1417 |
|
XM_017006215.1 |
XP_016861704.1
|
4, 5 |
446 |
|
XM_017006216.1 |
XP_016861705.1
|
3, 4 |
396 |
|
XM_017006217.1 |
XP_016861706.1
|
5, 6 |
493 |
|
XM_017006218.1 |
XP_016861707.1
|
3, 4 |
296 |
|
XM_017006220.1 |
XP_016861709.1
|
5, 6 |
461 |
|
XM_017006221.1 |
XP_016861710.1
|
3, 4 |
407 |
|
XM_017006222.1 |
XP_016861711.1
|
4, 5 |
455 |
|
XM_017006223.1 |
XP_016861712.1
|
5, 6 |
461 |
|
GenBank mRNA |
AK302040.1 |
- |
5, 6 |
444 |
|
AK302080.1 |
- |
4, 5 |
351 |
|
AK310249.1 |
- |
4, 5 |
380 |
|
AL133666.1 |
- |
5, 6 |
524 |
|
DC398430.1 |
- |
4, 5 |
399 |
|
View Full Product Details