Gene Transcripts

Gene Symbol
YIPF6
Entrez Gene ID
286451
Gene Name
Yip1 domain family member 6
Gene Aliases
FinGER6
Chromosome Location
Chr. X: 68498374 - 68572413 on Build GRCh38
UniGene ID
Hs.82719
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195214.1 NP_001182143.1 5 657
NM_173834.3 NP_776195.2 6 786
XM_011530923.2 XP_011529225.1 6 1194
XM_011530924.1 XP_011529226.1 6 568
XM_017029450.1 XP_016884939.1 6 568
XM_017029451.1 XP_016884940.1 5 1065
GenBank mRNA AK024673.1 - 2 92
AK292835.1 - 6 528
AK303992.1 - 5 387
BC012469.1 - 6 524 4452699

View Full Product Details