Gene Transcripts

Gene Symbol
HIC1
Entrez Gene ID
3090
Gene Name
hypermethylated in cancer 1
Gene Aliases
ZBTB29, ZNF901, hic-1
Chromosome Location
Chr. 17: 2055099 - 2059687 on Build GRCh38
UniGene ID
Hs.695682
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098202.1 NP_001091672.1 2 2188
NM_006497.3 NP_006488.2 2 2291

View Full Product Details