Gene Transcripts

Gene Symbol
C12orf74
Entrez Gene ID
338809
Gene Name
chromosome 12 open reading frame 74
Gene Aliases
Chromosome Location
Chr. 12: 92702843 - 92708549 on Build GRCh38
UniGene ID
Hs.326303
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037671.3 NP_001032760.1 2 915
NM_001178097.2 NP_001171568.1 2 915
GenBank mRNA BC043363.1 - 2 730
BC043364.1 - 2 730
BC105728.1 - 2 695 40025557
BC107813.1 - 2 694 40025555

View Full Product Details