Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178543.4 | NP_848638.3 | 3 | 817 | |
XM_011524737.1 | XP_011523039.1 | 3 | 1001 | ||
GenBank mRNA | AK126250.1 | - | 3 | 1001 | |
AY230663.1 | - | 3 | 800 | ||
AY358622.1 | - | 3 | 783 | ||
BC041453.1 | - | 3 | 800 | 5186743 |