Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042575.2 | NP_001036040.2 | 11, 12 | 1465 | |
NR_026734.1 | 12, 13 | 1578 | |||
XM_011512754.1 | XP_011511056.1 | 12, 13 | 1910 | ||
XM_011512758.1 | XP_011511060.1 | 11, 12 | 1331 | ||
GenBank mRNA | AK131211.1 | - | 12, 13 | 1578 | |
BC117322.1 | - | 11, 12 | 1377 | 40125873 |