Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000628.4 | NP_000619.3 | 3 | 329 | |
GenBank mRNA | AK124057.1 | - | 2 | 640 | |
AK299961.1 | - | 3 | 230 | ||
AK313348.1 | - | 3 | 322 | ||
AU133530.2 | - | 3 | 329 | ||
BC001903.1 | - | 3 | 308 | 3544611 | |
BT009777.1 | - | 3 | 223 | ||
JN591378.1 | - | 3 | 223 | ||
Z17227.1 | - | 3 | 266 |