Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001732.3 | NP_001001732.2 | 4 | 478 | |
NM_001159747.1 | NP_001153219.1 | 5 | 511 | ||
XM_011539789.2 | XP_011538091.1 | 12 | 1572 | ||
XM_011539790.2 | XP_011538092.1 | 12 | 1572 | ||
XM_011539791.2 | XP_011538093.1 | 11 | 1389 | ||
XM_011539792.2 | XP_011538094.1 | 6 | 694 | ||
XM_011539793.2 | XP_011538095.1 | 5 | 1673 | ||
XM_011539795.2 | XP_011538097.1 | 4 | 1490 | ||
XM_017016240.1 | XP_016871729.1 | 5 | 750 | ||
GenBank mRNA | AK302377.1 | - | 5 | 490 | |
BC075861.1 | - | 4 | 443 | 5171414 |