Gene Transcripts

Gene Symbol
MIR99AHG
Entrez Gene ID
388815
Gene Name
mir-99a-let-7c cluster host gene
Gene Aliases
C21orf34, C21orf35, LINC00478, MONC
Chromosome Location
Chr. 21: 16070488 - 16631727 on Build GRCh38
UniGene ID
Hs.473394
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027791.3 1 109
NR_136543.1 1 109
GenBank mRNA AL700268.1 - 1 20
CF135136.1 - 1 116
CF135139.1 - 1 116

View Full Product Details