Gene Transcripts

Gene Symbol
RGS7BP
Entrez Gene ID
401190
Gene Name
regulator of G-protein signaling 7 binding protein
Gene Aliases
R7BP
Chromosome Location
Chr. 5: 64504565 - 64614661 on Build GRCh38
UniGene ID
Hs.657133
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029875.2 NP_001025046.1 2 883
XM_005248502.3 XP_005248559.1 2 2265
GenBank mRNA AK296443.1 - 2 531
BC140707.1 - 2 229 9021575
BX640900.1 - 2 121

View Full Product Details