Gene Transcripts

Gene Symbol
MFAP1
Entrez Gene ID
4236
Gene Name
microfibrillar associated protein 1
Gene Aliases
Chromosome Location
Chr. 15: 43804535 - 43824753 on Build GRCh38
UniGene ID
Hs.61418
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005926.2 NP_005917.2 7 1146
GenBank mRNA AK312867.1 - 7 1122
BC023557.2 - 7 1081 4139958
BC050742.1 - 7 1092 6200464
U04209.1 - 7 1080

View Full Product Details