Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019598.2 | NP_062544.1 | 3 | 201 | |
NM_145894.1 | NP_665901.1 | 3 | 201 | ||
NM_145895.1 | NP_665902.1 | 3 | 201 | ||
XM_005258950.3 | XP_005259007.1 | 3 | 1109 | ||
XM_005258951.3 | XP_005259008.1 | 3 | 946 | ||
GenBank mRNA | AY358524.1 | - | 2 | 204 | |
BC035385.1 | - | 3 | 688 | 4825490 | |
BC136341.1 | - | 3 | 160 | 9020328 | |
BC143988.1 | - | 3 | 320 | ||
BC143990.1 | - | 3 | 160 | ||
BC143991.1 | - | 3 | 160 | ||
BX648580.1 | - | 1 | 112 |