Gene Transcripts

Gene Symbol
MXRA7
Entrez Gene ID
439921
Gene Name
matrix remodeling associated 7
Gene Aliases
PS1TP1, TMAP1
Chromosome Location
Chr. 17: 76673650 - 76711005 on Build GRCh38
UniGene ID
Hs.250723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008528.2 NP_001008528.1 4 658
XM_005257382.2 XP_005257439.1 3 744
XM_011524830.1 XP_011523132.1 4 544
GenBank mRNA AK123486.1 - 1 662
CK300836.1 - 4 368

View Full Product Details