Gene Transcripts

Gene Symbol
NDUFB8
Entrez Gene ID
4714
Gene Name
NADH:ubiquinone oxidoreductase subunit B8
Gene Aliases
ASHI, CI-ASHI
Chromosome Location
Chr. 10: 100523729 - 100529923 on Build GRCh38
UniGene ID
Hs.523215
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284367.1 NP_001271296.1 5 763
NM_001284368.1 NP_001271297.1 5 462
NM_005004.3 NP_004995.1 5 553
GenBank mRNA AA782554.1 - 1 202
AF044958.1 - 5 503
AF115968.1 - 5 509
AI282606.1 - 1 175
AK289579.1 - 5 494
AL080056.1 - 5 513
BC000466.2 - 5 494 2822781
BC019276.1 - 5 494 2822781
BM008944.1 - 5 450
CF127298.1 - 5 492
CR533490.1 - 5 481

View Full Product Details