Gene Transcripts

Gene Symbol
REV1
Entrez Gene ID
51455
Gene Name
REV1, DNA directed polymerase
Gene Aliases
AIBP80, REV1L
Chromosome Location
Chr. 2: 99400475 - 99490123 on Build GRCh38
UniGene ID
Hs.443077
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037872.2 NP_001032961.1 21 3773
NM_001321454.1 NP_001308383.1 22 3884
NM_001321455.1 NP_001308384.1 22 3930
NM_001321458.1 NP_001308387.1 22 3900
NM_001321459.1 NP_001308388.1 22 3976
NM_001321460.1 NP_001308389.1 22 3973
NM_016316.3 NP_057400.1 21 3776
NR_135649.1 22 3973
NR_135650.1 22 3930
NR_135651.1 20 3607
NR_135652.1 21 3776
NR_135653.1 20 3610
XM_011511341.2 XP_011509643.1 15 2393
XM_017004309.1 XP_016859798.1 22 4297
XM_017004310.1 XP_016859799.1 22 3663
XM_017004311.1 XP_016859800.1 22 3663
XM_017004312.1 XP_016859801.1 22 3876
XM_017004313.1 XP_016859802.1 21 5502
XM_017004314.1 XP_016859803.1 18 3153
XM_017004315.1 XP_016859804.1 18 3137
GenBank mRNA AB047646.1 - 21 3698
AF151538.1 - 21 3729
AF206019.1 - 21 3738
AF357886.1 - 21 3694
AJ131720.1 - 13 2095
BC037734.1 - 22 3911 4824689
BC130411.1 - 20 3544 40146442
CR736537.1 - 2 302

View Full Product Details