Gene Transcripts
- Gene Symbol
- STK26
- Entrez Gene ID
- 51765
- Gene Name
- serine/threonine protein kinase 26
- Gene Aliases
- MASK, MST4
- Chromosome Location
- Chr. X: 132023217 - 132075943 on Build GRCh38
- UniGene ID
- Hs.444247
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042452.1 |
NP_001035917.1
|
8, 9 |
905 |
|
NM_001042453.1 |
NP_001035918.1
|
9, 10 |
1151 |
|
NM_016542.3 |
NP_057626.2
|
10, 11 |
1382 |
|
XM_011531349.1 |
XP_011529651.1
|
11, 12 |
1355 |
|
XM_011531350.1 |
XP_011529652.1
|
10, 11 |
1428 |
|
GenBank mRNA |
AB040057.2 |
- |
10, 11 |
1301 |
|
AF231012.1 |
- |
10, 11 |
1270 |
|
AF344882.1 |
- |
9, 10 |
1091 |
|
AF344883.1 |
- |
8, 9 |
905 |
|
AK000556.1 |
- |
6, 7 |
714 |
|
AK027589.1 |
- |
5, 6 |
599 |
|
AK074837.1 |
- |
5, 6 |
635 |
|
AK075107.1 |
- |
9, 10 |
1063 |
|
AK303583.1 |
- |
11, 12 |
1363 |
|
AK314356.1 |
- |
10, 11 |
1298 |
|
AK316137.1 |
- |
11, 12 |
1363 |
|
BC098315.1 |
- |
10, 11 |
1287 |
40026349 |
BC099843.1 |
- |
10, 11 |
1287 |
40026348 |
BC103503.1 |
- |
10, 11 |
1287 |
40026350 |
BT020099.1 |
- |
9, 10 |
1081 |
|
View Full Product Details