Gene Transcripts
- Gene Symbol
- PLXNB3
- Entrez Gene ID
- 5365
- Gene Name
- plexin B3
- Gene Aliases
- PLEXB3, PLEXR, PLXN6
- Chromosome Location
- Chr. X: 153764196 - 153779346 on Build GRCh38
- UniGene ID
- Hs.632833
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163257.1 |
NP_001156729.1
|
14, 15 |
2683 |
|
NM_005393.2 |
NP_005384.2
|
13, 14 |
2517 |
|
GenBank mRNA |
AB033032.1 |
- |
13, 14 |
2457 |
|
AF149019.1 |
- |
13, 14 |
2457 |
|
AK292535.1 |
- |
13, 14 |
2517 |
|
AK295762.1 |
- |
13, 14 |
2537 |
|
AK295849.1 |
- |
10, 11 |
1207 |
|
AK295894.1 |
- |
12, 13 |
1401 |
|
AK304705.1 |
- |
13, 14 |
1582 |
|
AK307894.1 |
- |
14, 15 |
2660 |
|
View Full Product Details