Gene Transcripts

Gene Symbol
YTHDF1
Entrez Gene ID
54915
Gene Name
YTH N6-methyladenosine RNA binding protein 1
Gene Aliases
C20orf21
Chromosome Location
Chr. 20: 63195429 - 63216186 on Build GRCh38
UniGene ID
Hs.744852
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017798.3 NP_060268.2 4 1822
GenBank mRNA AB055518.1 - 1 1163
AK000398.1 - 2 983
AK095795.1 - 4 1540
AK300073.1 - 2 1552
AL834366.1 - 1 943
BC003681.1 - 3 1496
BC016920.2 - 1 1239
BC025264.1 - 1 1359
BC050284.1 - 4 1822 5298438

View Full Product Details