Gene Transcripts
- Gene Symbol
- RASIP1
- Entrez Gene ID
- 54922
- Gene Name
- Ras interacting protein 1
- Gene Aliases
- RAIN
- Chromosome Location
- Chr. 19: 48720585 - 48740833 on Build GRCh38
- UniGene ID
- Hs.233955
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017805.2 |
NP_060275.2
|
10, 11 |
2749 |
|
XM_011527053.2 |
XP_011525355.2
|
10, 11 |
2869 |
|
XM_011527054.2 |
XP_011525356.2
|
10, 11 |
2869 |
|
XM_011527055.2 |
XP_011525357.2
|
7, 8 |
2575 |
|
XM_017026914.1 |
XP_016882403.1
|
10, 11 |
2869 |
|
XM_017026915.1 |
XP_016882404.1
|
7, 8 |
2575 |
|
GenBank mRNA |
AK000408.1 |
- |
6, 7 |
864 |
|
AK123179.1 |
- |
2, 3 |
1046 |
|
AY378097.1 |
- |
10, 11 |
2614 |
|
BC021860.1 |
- |
6, 7 |
879 |
|
BC028614.1 |
- |
10, 11 |
2621 |
4815880 |
BX389914.2 |
- |
2, 3 |
248 |
|
View Full Product Details