Gene Transcripts
- Gene Symbol
- ANO10
- Entrez Gene ID
- 55129
- Gene Name
- anoctamin 10
- Gene Aliases
- SCAR10, TMEM16K
- Chromosome Location
- Chr. 3: 43366324 - 43691594 on Build GRCh38
- UniGene ID
- Hs.656657
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001204832.1 |
NP_001191761.1
|
10, 11 |
1781 |
|
NM_001204833.1 |
NP_001191762.1
|
9, 10 |
1646 |
|
NM_001204834.1 |
NP_001191763.1
|
10, 11 |
1409 |
|
NM_018075.3 |
NP_060545.3
|
11, 12 |
1979 |
|
XM_011533885.2 |
XP_011532187.2
|
13, 14 |
2166 |
|
XM_017006716.1 |
XP_016862205.1
|
12, 13 |
2062 |
|
XM_017006717.1 |
XP_016862206.1
|
12, 13 |
2014 |
|
XM_017006718.1 |
XP_016862207.1
|
12, 13 |
2028 |
|
XM_017006719.1 |
XP_016862208.1
|
11, 12 |
1816 |
|
XM_017006721.1 |
XP_016862210.1
|
10, 11 |
1708 |
|
XM_017006722.1 |
XP_016862211.1
|
10, 11 |
1681 |
|
GenBank mRNA |
AK096302.1 |
- |
11, 12 |
1979 |
|
AK131223.1 |
- |
10, 11 |
1697 |
|
AK292368.1 |
- |
11, 12 |
1864 |
|
AK295969.1 |
- |
9, 10 |
1581 |
|
AL832508.1 |
- |
11, 12 |
1846 |
|
BC038855.1 |
- |
10, 11 |
1307 |
|
View Full Product Details