Gene Transcripts

Gene Symbol
SBNO1
Entrez Gene ID
55206
Gene Name
strawberry notch homolog 1
Gene Aliases
MOP3, Sno
Chromosome Location
Chr. 12: 123289109 - 123365204 on Build GRCh38
UniGene ID
Hs.7012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167856.1 NP_001161328.1 3 462
NM_018183.3 NP_060653.3 3 459
XM_005253572.3 XP_005253629.1 4 600
XM_005253573.4 XP_005253630.1 5 1010
XM_005253575.3 XP_005253632.1 4 597
XM_005253576.3 XP_005253633.1 5 641
XM_006719473.3 XP_006719536.1 6 766
XM_006719474.3 XP_006719537.1 5 719
XM_011538533.2 XP_011536835.1 6 760
XM_011538534.2 XP_011536836.1 3 498
XM_011538535.2 XP_011536837.1 3 500
XM_017019556.1 XP_016875045.1 4 584
XM_017019557.1 XP_016875046.1 4 591
XM_017019558.1 XP_016875047.1 5 654
XM_017019559.1 XP_016875048.1 4 721
GenBank mRNA AB014772.1 - 3 459
AK001695.1 - 4 597
AK122703.1 - 5 1015
AK307488.1 - 3 498
AY364255.1 - 3 461
BC030544.1 - 4 593
BC133704.1 - 3 462 40124902

View Full Product Details