Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018404.2 | NP_060874.1 | 2, 3 | 495 | |
GenBank mRNA | AJ238994.1 | - | 2, 3 | 504 | |
AJ242782.1 | - | 2, 3 | 509 | ||
AJ272195.1 | - | 2, 3 | 316 | ||
AK074196.1 | - | 2, 3 | 552 | ||
AK074204.1 | - | 2, 3 | 323 | ||
AK222526.1 | - | 2, 3 | 323 | ||
BC033758.1 | - | 2, 3 | 311 | 5214358 |