Gene Transcripts

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
Chromosome Location
Chr. 5: 141355025 - 141512979 on Build GRCh38
UniGene ID
Hs.368160
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018917.3 NP_061740.2 1 1490
NM_032053.2 NP_114442.2 1 1490
GenBank mRNA AF152324.1 - 1 1314
AF152511.1 - 1 1314

View Full Product Details