Gene Transcripts

Gene Symbol
CHST7
Entrez Gene ID
56548
Gene Name
carbohydrate sulfotransferase 7
Gene Aliases
C6ST-2, GST-5
Chromosome Location
Chr. X: 46573687 - 46598496 on Build GRCh38
UniGene ID
Hs.129955
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019886.3 NP_063939.2 1 596
GenBank mRNA AB037187.1 - 1 361
AB040711.1 - 1 364
BC045537.1 - 1 499 5261787
BQ940898.1 - 1 526

View Full Product Details