Gene Transcripts
- Gene Symbol
- DIABLO
- Entrez Gene ID
- 56616
- Gene Name
- diablo IAP-binding mitochondrial protein
- Gene Aliases
- DFNA64, SMAC
- Chromosome Location
- Chr. 12: 122207662 - 122227534 on Build GRCh38
- UniGene ID
- Hs.169611
- Species
- Homo sapiens
- Gene Symbol
- B3GNT4
- Entrez Gene ID
- 79369
- Gene Name
- UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4
- Gene Aliases
- B3GN-T4, beta3Gn-T4
- Chromosome Location
- Chr. 12: 122203681 - 122208135 on Build GRCh38
- UniGene ID
- Hs.363315
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095746.1 |
- |
2 |
2259 |
|
View Full Product Details