Gene Transcripts

Gene Symbol
PSMB1
Entrez Gene ID
5689
Gene Name
proteasome subunit beta 1
Gene Aliases
HC5, PMSB1, PSC5
Chromosome Location
Chr. 6: 170535116 - 170553329 on Build GRCh38
UniGene ID
Hs.352768
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002793.3 NP_002784.1 6 787
GenBank mRNA AB451312.1 - 6 700
AB451442.1 - 6 700
AK023290.1 - 5 1915
AK058050.1 - 6 757
AK223193.1 - 6 738
BC000508.2 - 6 738 2822268
BC020807.1 - 6 738 4281426
BQ685374.1 - 5 517
BT019720.1 - 6 700
D00761.1 - 6 712

View Full Product Details