Gene Transcripts

Gene Symbol
ZNF77
Entrez Gene ID
58492
Gene Name
zinc finger protein 77
Gene Aliases
pT1
Chromosome Location
Chr. 19: 2933218 - 2944971 on Build GRCh38
UniGene ID
Hs.38004
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021217.2 NP_067040.1 2, 3 261
XM_017027082.1 XP_016882571.1 2, 3 286
GenBank mRNA BC043354.1 - 2, 3 222
BC062561.1 - 2, 3 223 6644579

View Full Product Details