Gene Transcripts
- Gene Symbol
- RAB1A
- Entrez Gene ID
- 5861
- Gene Name
- RAB1A, member RAS oncogene family
- Gene Aliases
- RAB1, YPT1
- Chromosome Location
- Chr. 2: 65086854 - 65130301 on Build GRCh38
- UniGene ID
- Hs.310645
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004161.4 |
NP_004152.1
|
2, 3 |
474 |
|
NM_015543.1 |
NP_056358.1
|
2, 3 |
474 |
|
XM_005264468.1 |
XP_005264525.1
|
2, 3 |
279 |
|
GenBank mRNA |
AF498929.1 |
- |
2, 3 |
88 |
|
AK303678.1 |
- |
2, 3 |
278 |
|
AK315495.1 |
- |
2, 3 |
283 |
|
AL050268.1 |
- |
2, 3 |
278 |
|
BC000905.2 |
- |
2, 3 |
266 |
2900705 |
BG704216.1 |
- |
2, 3 |
588 |
|
BT019528.1 |
- |
2, 3 |
88 |
|
BT019529.1 |
- |
2, 3 |
88 |
|
BX571747.1 |
- |
2, 3 |
271 |
|
CR533479.1 |
- |
2, 3 |
88 |
|
CR536488.1 |
- |
2, 3 |
88 |
|
M28209.1 |
- |
2, 3 |
138 |
|
X13898.1 |
- |
2, 3 |
88 |
|
View Full Product Details