Gene Transcripts

Gene Symbol
EDDM3B
Entrez Gene ID
64184
Gene Name
epididymal protein 3B
Gene Aliases
EP3B, FAM12B, HE3-BETA, HE3B, RAM2
Chromosome Location
Chr. 14: 20768427 - 20770948 on Build GRCh38
UniGene ID
Hs.525202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022360.4 NP_071755.1 2 155
GenBank mRNA AY358865.1 - 2 155
BC128030.1 - 2 118 40002822
BX112699.1 - 2 151
X76386.2 - 1 117

View Full Product Details