Gene Transcripts

Gene Symbol
GMNC
Entrez Gene ID
647309
Gene Name
geminin coiled-coil domain containing
Gene Aliases
GEMC1
Chromosome Location
Chr. 3: 190843271 - 190864200 on Build GRCh38
UniGene ID
Hs.518490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146686.2 NP_001140158.1 5 743
GenBank mRNA BC031680.1 - 1 488
CR749823.1 - 2 576

View Full Product Details