Gene Transcripts

Gene Symbol
CERK
Entrez Gene ID
64781
Gene Name
ceramide kinase
Gene Aliases
LK4, dA59H18.2, dA59H18.3, hCERK
Chromosome Location
Chr. 22: 46684410 - 46738255 on Build GRCh38
UniGene ID
Hs.200668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022766.5 NP_073603.2 4 503
XM_017028909.1 XP_016884398.1 4 300
GenBank mRNA AB051433.1 - 3 229
AB079066.1 - 4 519
AJ457828.1 - 4 466
AK291476.1 - 4 459
BC067255.1 - 4 455
BC126940.1 - 4 446 40084110
CR456404.1 - 4 406

View Full Product Details