Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001011554.2 | NP_001011554.1 | 11 | 1350 | |
NM_001193339.1 | NP_001180268.1 | 9 | 1164 | ||
NM_001193340.1 | NP_001180269.1 | 10 | 1245 | ||
NM_001193342.1 | NP_001180271.1 | 10 | 1141 | ||
NM_022829.5 | NP_073740.2 | 10 | 1314 | ||
GenBank mRNA | AF154121.1 | - | 10 | 1293 | |
AK096658.1 | - | 10 | 1245 | ||
AK293902.1 | - | 10 | 1130 | ||
AK310960.1 | - | 9 | 1324 | ||
AK315131.1 | - | 10 | 1312 | ||
AY072810.3 | - | 10 | 1293 | ||
BC035966.1 | - | 10 | 1312 | 4609776 | |
DA272524.1 | - | 5 | 491 |