Gene Transcripts

Gene Symbol
CT47B1
Entrez Gene ID
643311
Gene Name
cancer/testis antigen family 47, member B1
Gene Aliases
CT47.13, CT47A13
Chromosome Location
Chr. X: 120872554 - 120875925 on Build GRCh38
UniGene ID
Hs.729566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145718.1 NP_001139190.1 1 535
XM_017029734.1 XP_016885223.1 1 497

View Full Product Details