Gene Transcripts

Gene Symbol
YWHAH
Entrez Gene ID
7533
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein eta
Gene Aliases
YWHA1
Chromosome Location
Chr. 22: 31944493 - 31957603 on Build GRCh38
UniGene ID
Hs.226755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003405.3 NP_003396.1 2 878
GenBank mRNA AK307750.1 - 3 1002
AK312650.1 - 2 722
AY007132.1 - 1 465
BC003047.1 - 2 813 3543571
CR456612.1 - 2 681
L20422.1 - 2 646
S80794.1 - 2 828
X57345.1 - 1 561
X78138.1 - 2 837

View Full Product Details