Gene Transcripts

Gene Symbol
ST8SIA4
Entrez Gene ID
7903
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4
Gene Aliases
PST, PST1, SIAT8D, ST8SIA-IV
Chromosome Location
Chr. 5: 100806935 - 100903285 on Build GRCh38
UniGene ID
Hs.308628
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005668.5 NP_005659.1 3 735
NM_175052.2 NP_778222.1 3 735
XM_005272078.2 XP_005272135.1 3 735
XM_011543630.1 XP_011541932.1 3 735
GenBank mRNA AK292872.1 - 3 687
BC027866.1 - 3 664
BC053657.1 - 3 716 5752337
BM921020.1 - 3 735
L41680.1 - 3 617

View Full Product Details