Gene Transcripts

Gene Symbol
BAIAP2L2
Entrez Gene ID
80115
Gene Name
BAI1 associated protein 2 like 2
Gene Aliases
Chromosome Location
Chr. 22: 38084889 - 38110944 on Build GRCh38
UniGene ID
Hs.474822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025045.5 NP_079321.3 7 667
XM_005261751.4 XP_005261808.1 8 668
XM_011530379.2 XP_011528681.1 7 784
XM_011530380.2 XP_011528682.1 6 570
XM_011530381.2 XP_011528683.1 7 561
XM_011530382.2 XP_011528684.1 7 949
XM_011530383.2 XP_011528685.1 6 887
XM_011530384.2 XP_011528686.1 7 825
GenBank mRNA AK026235.1 - 7 625
AY358164.1 - 7 667

View Full Product Details