Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015981.3 | NP_057065.2 | 9 | 892 | |
NM_171825.2 | NP_741960.1 | 9 | 892 | ||
XM_017009898.1 | XP_016865387.1 | 9 | 892 | ||
GenBank mRNA | AB023185.1 | - | 9 | 825 | |
AF091486.1 | - | 9 | 781 | ||
AF145710.1 | - | 9 | 683 | ||
AF145711.1 | - | 9 | 683 | ||
AK289776.1 | - | 9 | 892 | ||
AK308853.1 | - | 8 | 765 | ||
AY429524.1 | - | 3 | 184 |