Gene Transcripts
- Gene Symbol
- FYTTD1
- Entrez Gene ID
- 84248
- Gene Name
- forty-two-three domain containing 1
- Gene Aliases
- UIF
- Chromosome Location
- Chr. 3: 197749553 - 197784446 on Build GRCh38
- UniGene ID
- Hs.277533
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001011537.2 |
NP_001011537.2
|
4, 5 |
534 |
|
NM_032288.6 |
NP_115664.2
|
3, 4 |
593 |
|
NR_027840.1 |
|
3, 4 |
704 |
|
GenBank mRNA |
AJ344094.1 |
- |
3, 4 |
439 |
|
AK027672.1 |
- |
3, 4 |
477 |
|
AK302546.1 |
- |
3, 4 |
704 |
|
AK303462.1 |
- |
4, 5 |
534 |
|
AK315019.1 |
- |
3, 4 |
442 |
|
AK316308.1 |
- |
3, 4 |
334 |
|
AL136558.1 |
- |
3, 4 |
446 |
|
BC035006.2 |
- |
3, 4 |
596 |
5267824 |
BC039734.1 |
- |
3, 4 |
503 |
6042926 |
BX509179.1 |
- |
4, 5 |
617 |
|
CR533502.1 |
- |
3, 4 |
374 |
|
View Full Product Details