Gene Transcripts

Gene Symbol
WDR83
Entrez Gene ID
84292
Gene Name
WD repeat domain 83
Gene Aliases
MORG1
Chromosome Location
Chr. 19: 12666804 - 12675832 on Build GRCh38
UniGene ID
Hs.657204
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099737.2 NP_001093207.1 3, 4 444
NM_032332.3 NP_115708.1 1, 2 187
NR_029375.1 3, 4 444
GenBank mRNA AK074525.1 - 3, 4 434
AK223195.1 - 1, 2 137
AK314164.1 - 3, 4 421
BC005870.2 - 1, 2 120 2961616
CB143206.1 - 1, 2 187

View Full Product Details