Gene Transcripts

Gene Symbol
SFXN5
Entrez Gene ID
94097
Gene Name
sideroflexin 5
Gene Aliases
BBG-TCC
Chromosome Location
Chr. 2: 72942036 - 73072234 on Build GRCh38
UniGene ID
Hs.368171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144579.2 NP_653180.1 1, 2 231
XM_005264648.1 XP_005264705.1 1, 2 137
XM_006712147.1 XP_006712210.1 1, 2 137
XM_011533189.1 XP_011531491.1 1, 2 137
XM_011533190.2 XP_011531492.1 1, 2 629
XM_011533197.1 XP_011531499.1 1, 2 137
XM_017005342.1 XP_016860831.1 1, 2 137
XM_017005346.1 XP_016860835.1 1, 2 104
GenBank mRNA AK289731.1 - 1, 2 120
AK314770.1 - 1, 2 119
AL833454.1 - 1, 2 105
AY044437.1 - 1, 2 235
BC101311.2 - 1, 2 231 40024811
BC101312.1 - 1, 2 231 40024812
BC101313.2 - 1, 2 231 40024813

View Full Product Details