Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014763.3 | NP_055578.2 | 2, 3 | 254 | |
GenBank mRNA | AK291250.1 | - | 2, 3 | 240 | |
AK295578.1 | - | 2, 3 | 226 | ||
BC030144.2 | - | 2, 3 | 239 | 4763405 | |
BI551514.1 | - | 2, 3 | 259 | ||
BQ432684.1 | - | 2, 3 | 257 | ||
D14660.1 | - | 2, 3 | 227 |