Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113202301-113202805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAM1
Entrez Gene ID
4684
Gene Name
neural cell adhesion molecule 1
Gene Aliases
CD56, MSK39, NCAM
UniGene
Hs.503878
Gene Chromosome Location
Chr.11: 112961247-113278436 on Build GRCh38
Transcripts
NM_000615.6 NM_001076682.3 NM_001242607.1 NM_001242608.1 NM_181351.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3443424 c.55G>A p.G19R Chr.11: 113202411-113202411 on Build GRCh38
1666493 c.25-10_25-9insTTTG p.? Chr.11: 113202344-113202345 on Build GRCh38
1216762 c.84C>A p.F28L Chr.11: 113202440-113202440 on Build GRCh38
5076305 c.51T>G p.V17V Chr.11: 113202380-113202380 on Build GRCh38
3443421 c.37G>A p.D13N Chr.11: 113202393-113202393 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562067652 Chr.11: 113202664-113202664 on Build GRCh38
rs45561332 Chr.11: 113202449-113202449 on Build GRCh38
rs781966417 Chr.11: 113202364-113202364 on Build GRCh38
rs781994544 Chr.11: 113202469-113202469 on Build GRCh38
rs782220003 Chr.11: 113202410-113202410 on Build GRCh38
rs782225091 Chr.11: 113202446-113202446 on Build GRCh38
rs546072497 Chr.11: 113202502-113202502 on Build GRCh38
rs112087457 Chr.11: 113202332-113202333 on Build GRCh38
rs782393320 Chr.11: 113202369-113202370 on Build GRCh38
rs782759996 Chr.11: 113202329-113202330 on Build GRCh38
rs782593754 Chr.11: 113202423-113202423 on Build GRCh38
rs781986082 Chr.11: 113202371-113202372 on Build GRCh38
rs377030084 Chr.11: 113202399-113202399 on Build GRCh38
rs781942589 Chr.11: 113202372-113202373 on Build GRCh38
rs199912330 Chr.11: 113202342-113202342 on Build GRCh38
rs782696265 Chr.11: 113202380-113202380 on Build GRCh38
rs563239907 Chr.11: 113202349-113202349 on Build GRCh38
rs782776253 Chr.11: 113202342-113202343 on Build GRCh38
rs886074140 Chr.11: 113202581-113202581 on Build GRCh38
rs1045074399 Chr.11: 113202754-113202754 on Build GRCh38
rs201978903 Chr.11: 113202330-113202330 on Build GRCh38
rs781907852 Chr.11: 113202331-113202332 on Build GRCh38
rs975094581 Chr.11: 113202548-113202548 on Build GRCh38
rs60566134 Chr.11: 113202373-113202374 on Build GRCh38
rs782636804 Chr.11: 113202637-113202637 on Build GRCh38
rs782773690 Chr.11: 113202374-113202374 on Build GRCh38
rs781886879 Chr.11: 113202376-113202376 on Build GRCh38
rs373994214 Chr.11: 113202398-113202398 on Build GRCh38
rs782742927 Chr.11: 113202359-113202359 on Build GRCh38
rs782162099 Chr.11: 113202377-113202377 on Build GRCh38
rs782471027 Chr.11: 113202384-113202384 on Build GRCh38
rs782223470 Chr.11: 113202331-113202331 on Build GRCh38
rs369951427 Chr.11: 113202543-113202543 on Build GRCh38
rs782058571 Chr.11: 113202354-113202354 on Build GRCh38
rs45448394 Chr.11: 113202333-113202333 on Build GRCh38
rs559682484 Chr.11: 113202506-113202506 on Build GRCh38
rs200328067 Chr.11: 113202344-113202344 on Build GRCh38
rs782631468 Chr.11: 113202392-113202392 on Build GRCh38
rs72188360 Chr.11: 113202363-113202364 on Build GRCh38
rs782481856 Chr.11: 113202333-113202333 on Build GRCh38
rs933029630 Chr.11: 113202566-113202566 on Build GRCh38
rs1836796 Chr.11: 113202638-113202638 on Build GRCh38
rs781808973 Chr.11: 113202382-113202382 on Build GRCh38
rs1003086908 Chr.11: 113202766-113202766 on Build GRCh38
rs782110765 Chr.11: 113202369-113202369 on Build GRCh38
rs577218059 Chr.11: 113202406-113202406 on Build GRCh38
rs781916691 Chr.11: 113202351-113202351 on Build GRCh38
rs1037430916 Chr.11: 113202663-113202663 on Build GRCh38
rs528313460 Chr.11: 113202563-113202563 on Build GRCh38
rs370251266 Chr.11: 113202495-113202495 on Build GRCh38
rs782585196 Chr.11: 113202328-113202328 on Build GRCh38
rs564603741 Chr.11: 113202542-113202542 on Build GRCh38
rs782145981 Chr.11: 113202477-113202477 on Build GRCh38
rs782322515 Chr.11: 113202409-113202409 on Build GRCh38
rs782363836 Chr.11: 113202455-113202455 on Build GRCh38
rs1051383124 Chr.11: 113202568-113202568 on Build GRCh38
rs914410154 Chr.11: 113202464-113202464 on Build GRCh38
rs76487552 Chr.11: 113202332-113202332 on Build GRCh38
rs782419904 Chr.11: 113202747-113202747 on Build GRCh38
rs5794855 Chr.11: 113202344-113202345 on Build GRCh38
rs921844953 Chr.11: 113202562-113202562 on Build GRCh38
rs549482336 Chr.11: 113202368-113202368 on Build GRCh38
rs782596452 Chr.11: 113202335-113202336 on Build GRCh38
rs906111707 Chr.11: 113202763-113202763 on Build GRCh38
rs561422423 Chr.11: 113202727-113202727 on Build GRCh38

View Full Product Details