Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113207177-113207680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAM1
Entrez Gene ID
4684
Gene Name
neural cell adhesion molecule 1
Gene Aliases
CD56, MSK39, NCAM
UniGene
Hs.503878
Gene Chromosome Location
Chr.11: 112961247-113278436 on Build GRCh38
Transcripts
NM_000615.6 NM_001076682.3 NM_001242607.1 NM_001242608.1 NM_181351.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6130940 c.672G>C p.V224V Chr.11: 113207334-113207334 on Build GRCh38
5416872 c.734G>A p.S245N Chr.11: 113207369-113207369 on Build GRCh38
5371655 c.666C>T p.T222T Chr.11: 113207328-113207328 on Build GRCh38
6217834 c.599-6C>A p.? Chr.11: 113207255-113207255 on Build GRCh38
3808358 c.664A>C p.T222P Chr.11: 113207326-113207326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045630917 Chr.11: 113207629-113207629 on Build GRCh38
rs782593612 Chr.11: 113207423-113207423 on Build GRCh38
rs782340784 Chr.11: 113207276-113207276 on Build GRCh38
rs141963340 Chr.11: 113207430-113207430 on Build GRCh38
rs782117392 Chr.11: 113207417-113207417 on Build GRCh38
rs554416377 Chr.11: 113207504-113207504 on Build GRCh38
rs371142479 Chr.11: 113207322-113207322 on Build GRCh38
rs782424337 Chr.11: 113207401-113207401 on Build GRCh38
rs781969531 Chr.11: 113207286-113207286 on Build GRCh38
rs964052195 Chr.11: 113207247-113207247 on Build GRCh38
rs782216041 Chr.11: 113207221-113207221 on Build GRCh38
rs782122142 Chr.11: 113207311-113207311 on Build GRCh38
rs544657433 Chr.11: 113207475-113207475 on Build GRCh38
rs781918930 Chr.11: 113207256-113207256 on Build GRCh38
rs782021360 Chr.11: 113207314-113207314 on Build GRCh38
rs782752474 Chr.11: 113207568-113207568 on Build GRCh38
rs575407443 Chr.11: 113207459-113207459 on Build GRCh38
rs941385226 Chr.11: 113207519-113207519 on Build GRCh38
rs782450123 Chr.11: 113207212-113207212 on Build GRCh38
rs146084574 Chr.11: 113207458-113207458 on Build GRCh38
rs782190874 Chr.11: 113207262-113207262 on Build GRCh38
rs915717393 Chr.11: 113207572-113207572 on Build GRCh38
rs1030095846 Chr.11: 113207465-113207465 on Build GRCh38
rs983304948 Chr.11: 113207492-113207492 on Build GRCh38
rs1031608349 Chr.11: 113207206-113207206 on Build GRCh38
rs17115154 Chr.11: 113207313-113207313 on Build GRCh38
rs782751481 Chr.11: 113207329-113207329 on Build GRCh38
rs781993227 Chr.11: 113207397-113207397 on Build GRCh38
rs376337341 Chr.11: 113207337-113207337 on Build GRCh38
rs563070008 Chr.11: 113207319-113207319 on Build GRCh38
rs372902478 Chr.11: 113207246-113207246 on Build GRCh38
rs551838234 Chr.11: 113207418-113207418 on Build GRCh38
rs782792136 Chr.11: 113207424-113207424 on Build GRCh38
rs575741314 Chr.11: 113207463-113207463 on Build GRCh38
rs199787286 Chr.11: 113207302-113207302 on Build GRCh38
rs782465505 Chr.11: 113207362-113207362 on Build GRCh38
rs782698294 Chr.11: 113207323-113207323 on Build GRCh38
rs186226516 Chr.11: 113207585-113207585 on Build GRCh38
rs75844619 Chr.11: 113207394-113207394 on Build GRCh38
rs973908530 Chr.11: 113207566-113207566 on Build GRCh38
rs955383628 Chr.11: 113207466-113207466 on Build GRCh38
rs975386483 Chr.11: 113207283-113207283 on Build GRCh38
rs571985408 Chr.11: 113207488-113207488 on Build GRCh38
rs371057906 Chr.11: 113207344-113207344 on Build GRCh38
rs17115156 Chr.11: 113207325-113207325 on Build GRCh38
rs782591967 Chr.11: 113207363-113207363 on Build GRCh38
rs782530698 Chr.11: 113207338-113207338 on Build GRCh38
rs180883651 Chr.11: 113207432-113207432 on Build GRCh38
rs199831399 Chr.11: 113207409-113207409 on Build GRCh38
rs782491247 Chr.11: 113207239-113207239 on Build GRCh38
rs906894322 Chr.11: 113207633-113207633 on Build GRCh38
rs782323015 Chr.11: 113207304-113207304 on Build GRCh38
rs782677309 Chr.11: 113207343-113207343 on Build GRCh38
rs148136990 Chr.11: 113207245-113207245 on Build GRCh38
rs1022381551 Chr.11: 113207457-113207457 on Build GRCh38
rs781973138 Chr.11: 113207413-113207413 on Build GRCh38
rs565989209 Chr.11: 113207398-113207398 on Build GRCh38
rs782468975 Chr.11: 113207327-113207327 on Build GRCh38
rs138849431 Chr.11: 113207476-113207476 on Build GRCh38
rs17115157 Chr.11: 113207393-113207393 on Build GRCh38
rs782071747 Chr.11: 113207402-113207402 on Build GRCh38
rs906842307 Chr.11: 113207408-113207408 on Build GRCh38
rs782409634 Chr.11: 113207252-113207252 on Build GRCh38

View Full Product Details