Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113214289-113214799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAM1
Entrez Gene ID
4684
Gene Name
neural cell adhesion molecule 1
Gene Aliases
CD56, MSK39, NCAM
UniGene
Hs.503878
Gene Chromosome Location
Chr.11: 112961247-113278436 on Build GRCh38
Transcripts
NM_000615.6 NM_001076682.3 NM_001242607.1 NM_001242608.1 NM_181351.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7278605 c.937G>C p.E313Q Chr.11: 113214392-113214392 on Build GRCh38
3443449 c.969G>A p.Q323Q Chr.11: 113214424-113214424 on Build GRCh38
3443452 c.1006C>T p.P336S Chr.11: 113214461-113214461 on Build GRCh38
1351310 c.1026T>G p.T342T Chr.11: 113214481-113214481 on Build GRCh38
7278603 c.910G>C p.E304Q Chr.11: 113214392-113214392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782534608 Chr.11: 113214482-113214482 on Build GRCh38
rs782648874 Chr.11: 113214368-113214368 on Build GRCh38
rs782718534 Chr.11: 113214547-113214547 on Build GRCh38
rs563453559 Chr.11: 113214727-113214727 on Build GRCh38
rs782440279 Chr.11: 113214339-113214339 on Build GRCh38
rs782276642 Chr.11: 113214369-113214369 on Build GRCh38
rs782124434 Chr.11: 113214425-113214425 on Build GRCh38
rs192718886 Chr.11: 113214317-113214317 on Build GRCh38
rs1051117750 Chr.11: 113214551-113214551 on Build GRCh38
rs969913624 Chr.11: 113214424-113214424 on Build GRCh38
rs568391383 Chr.11: 113214489-113214489 on Build GRCh38
rs572105065 Chr.11: 113214640-113214640 on Build GRCh38
rs185400485 Chr.11: 113214503-113214503 on Build GRCh38
rs968464370 Chr.11: 113214748-113214748 on Build GRCh38
rs373827595 Chr.11: 113214536-113214536 on Build GRCh38
rs548503538 Chr.11: 113214459-113214459 on Build GRCh38
rs782338616 Chr.11: 113214397-113214397 on Build GRCh38
rs373516826 Chr.11: 113214387-113214387 on Build GRCh38
rs781979983 Chr.11: 113214419-113214419 on Build GRCh38
rs781922415 Chr.11: 113214390-113214390 on Build GRCh38
rs576965258 Chr.11: 113214758-113214758 on Build GRCh38
rs962854345 Chr.11: 113214360-113214360 on Build GRCh38
rs528456293 Chr.11: 113214448-113214448 on Build GRCh38
rs140683750 Chr.11: 113214330-113214330 on Build GRCh38
rs201693511 Chr.11: 113214500-113214500 on Build GRCh38
rs974056776 Chr.11: 113214394-113214394 on Build GRCh38
rs782751987 Chr.11: 113214469-113214469 on Build GRCh38
rs370947557 Chr.11: 113214493-113214493 on Build GRCh38
rs371744296 Chr.11: 113214555-113214555 on Build GRCh38
rs782722769 Chr.11: 113214641-113214641 on Build GRCh38
rs782091615 Chr.11: 113214465-113214465 on Build GRCh38
rs35696834 Chr.11: 113214721-113214722 on Build GRCh38
rs781931393 Chr.11: 113214517-113214517 on Build GRCh38
rs995754576 Chr.11: 113214649-113214649 on Build GRCh38
rs781794964 Chr.11: 113214488-113214488 on Build GRCh38
rs932916664 Chr.11: 113214766-113214766 on Build GRCh38
rs376521254 Chr.11: 113214507-113214507 on Build GRCh38
rs376253720 Chr.11: 113214429-113214429 on Build GRCh38
rs763403735 Chr.11: 113214570-113214570 on Build GRCh38
rs686050 Chr.11: 113214522-113214522 on Build GRCh38
rs782025802 Chr.11: 113214436-113214436 on Build GRCh38
rs912604221 Chr.11: 113214775-113214775 on Build GRCh38
rs528399400 Chr.11: 113214723-113214723 on Build GRCh38
rs529317020 Chr.11: 113214676-113214676 on Build GRCh38
rs1002727454 Chr.11: 113214663-113214663 on Build GRCh38
rs1022767894 Chr.11: 113214657-113214657 on Build GRCh38
rs1051338232 Chr.11: 113214767-113214767 on Build GRCh38
rs955698814 Chr.11: 113214724-113214724 on Build GRCh38
rs2241449 Chr.11: 113214666-113214666 on Build GRCh38
rs782196262 Chr.11: 113214391-113214391 on Build GRCh38
rs781984390 Chr.11: 113214529-113214529 on Build GRCh38
rs782601339 Chr.11: 113214355-113214355 on Build GRCh38
rs782687432 Chr.11: 113214486-113214486 on Build GRCh38
rs554926510 Chr.11: 113214708-113214708 on Build GRCh38
rs1004332177 Chr.11: 113214598-113214598 on Build GRCh38
rs969967771 Chr.11: 113214658-113214658 on Build GRCh38
rs782502104 Chr.11: 113214365-113214365 on Build GRCh38
rs928280803 Chr.11: 113214487-113214487 on Build GRCh38
rs367870034 Chr.11: 113214544-113214544 on Build GRCh38
rs1023305830 Chr.11: 113214407-113214407 on Build GRCh38
rs686934 Chr.11: 113214726-113214726 on Build GRCh38
rs546316210 Chr.11: 113214764-113214764 on Build GRCh38
rs782127737 Chr.11: 113214530-113214530 on Build GRCh38
rs367708183 Chr.11: 113214638-113214638 on Build GRCh38
rs574751969 Chr.11: 113214709-113214709 on Build GRCh38
rs782074654 Chr.11: 113214521-113214521 on Build GRCh38
rs781814795 Chr.11: 113214463-113214463 on Build GRCh38
rs781874490 Chr.11: 113214471-113214471 on Build GRCh38
rs782230481 Chr.11: 113214359-113214359 on Build GRCh38
rs781905057 Chr.11: 113214540-113214540 on Build GRCh38

View Full Product Details