Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10637852-10638344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASZ1
Entrez Gene ID
54897
Gene Name
castor zinc finger 1
Gene Aliases
CAS11, CST, SRG, ZNF693, dJ734G22.1
UniGene
Hs.439894
Gene Chromosome Location
Chr.1: 10636602-10796676 on Build GRCh38
Transcripts
NM_001079843.2 XM_017001539.1 XM_017001540.1 XM_017001541.1

SNP Detail

SNP ID Chromosome Location
rs1038998583 Chr.1: 10638271-10638271 on Build GRCh38
rs570852916 Chr.1: 10638253-10638253 on Build GRCh38
rs191369987 Chr.1: 10638143-10638143 on Build GRCh38
rs912287875 Chr.1: 10638029-10638029 on Build GRCh38
rs373225643 Chr.1: 10638182-10638182 on Build GRCh38
rs187980089 Chr.1: 10638134-10638134 on Build GRCh38
rs148704981 Chr.1: 10638167-10638167 on Build GRCh38
rs1043764918 Chr.1: 10637945-10637945 on Build GRCh38
rs902136565 Chr.1: 10638272-10638272 on Build GRCh38
rs867290124 Chr.1: 10637927-10637927 on Build GRCh38
rs552312055 Chr.1: 10638230-10638230 on Build GRCh38
rs953698338 Chr.1: 10637957-10637957 on Build GRCh38
rs1018287920 Chr.1: 10638021-10638021 on Build GRCh38
rs906607056 Chr.1: 10638195-10638195 on Build GRCh38
rs985132423 Chr.1: 10637977-10637977 on Build GRCh38
rs943374905 Chr.1: 10638268-10638268 on Build GRCh38
rs1011826535 Chr.1: 10638267-10638267 on Build GRCh38
rs534779019 Chr.1: 10638285-10638285 on Build GRCh38
rs575046161 Chr.1: 10638112-10638121 on Build GRCh38
rs918792343 Chr.1: 10638055-10638055 on Build GRCh38
rs916821540 Chr.1: 10638141-10638141 on Build GRCh38
rs1046397547 Chr.1: 10638178-10638178 on Build GRCh38
rs571252771 Chr.1: 10638250-10638250 on Build GRCh38
rs972878747 Chr.1: 10638047-10638047 on Build GRCh38
rs928550684 Chr.1: 10638222-10638222 on Build GRCh38
rs1009602888 Chr.1: 10637935-10637935 on Build GRCh38
rs546365765 Chr.1: 10638202-10638202 on Build GRCh38
rs530366392 Chr.1: 10638124-10638124 on Build GRCh38
rs1048682166 Chr.1: 10638115-10638115 on Build GRCh38
rs938560064 Chr.1: 10638234-10638234 on Build GRCh38
rs191609779 Chr.1: 10637915-10637915 on Build GRCh38
rs371126309 Chr.1: 10638137-10638139 on Build GRCh38
rs553092606 Chr.1: 10638296-10638296 on Build GRCh38
rs903910155 Chr.1: 10638259-10638259 on Build GRCh38
rs182954626 Chr.1: 10637939-10637939 on Build GRCh38
rs777995850 Chr.1: 10638123-10638123 on Build GRCh38
rs956134924 Chr.1: 10637899-10637899 on Build GRCh38
rs559975152 Chr.1: 10638108-10638108 on Build GRCh38
rs965144616 Chr.1: 10638038-10638038 on Build GRCh38
rs546022463 Chr.1: 10638166-10638166 on Build GRCh38
rs1026482453 Chr.1: 10638101-10638101 on Build GRCh38
rs778773590 Chr.1: 10638056-10638056 on Build GRCh38
rs528149862 Chr.1: 10638075-10638075 on Build GRCh38
rs970146044 Chr.1: 10638176-10638176 on Build GRCh38
rs1052873334 Chr.1: 10638237-10638237 on Build GRCh38
rs903837652 Chr.1: 10637958-10637958 on Build GRCh38
rs1026513759 Chr.1: 10638313-10638313 on Build GRCh38
rs997690314 Chr.1: 10638056-10638056 on Build GRCh38
rs2624879 Chr.1: 10638306-10638306 on Build GRCh38
rs540361726 Chr.1: 10638077-10638077 on Build GRCh38
rs988642192 Chr.1: 10638240-10638240 on Build GRCh38
rs555684 Chr.1: 10638161-10638161 on Build GRCh38
rs564178454 Chr.1: 10638073-10638073 on Build GRCh38
rs886590091 Chr.1: 10638314-10638314 on Build GRCh38
rs564213932 Chr.1: 10638194-10638194 on Build GRCh38

View Full Product Details