Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11954751-11955256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLOD1
Entrez Gene ID
5351
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 1
Gene Aliases
EDS6, LH, LH1, LLH, PLOD
UniGene
Hs.75093
Gene Chromosome Location
Chr.1: 11934667-11975542 on Build GRCh38
Transcripts
NM_000302.3 NM_001316320.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7384617 c.608G>A p.R203H Chr.1: 11954858-11954858 on Build GRCh38
6750242 c.607C>T p.R203C Chr.1: 11954857-11954857 on Build GRCh38
7384618 c.749G>A p.R250H Chr.1: 11954858-11954858 on Build GRCh38
6335797 c.643+6C>T p.? Chr.1: 11954899-11954899 on Build GRCh38
7384619 c.608G>A p.R203H Chr.1: 11954858-11954858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940663229 Chr.1: 11954930-11954930 on Build GRCh38
rs950146670 Chr.1: 11955189-11955189 on Build GRCh38
rs145642241 Chr.1: 11955085-11955085 on Build GRCh38
rs201152076 Chr.1: 11954938-11954938 on Build GRCh38
rs1045586863 Chr.1: 11955200-11955200 on Build GRCh38
rs753770094 Chr.1: 11954937-11954937 on Build GRCh38
rs775042399 Chr.1: 11954961-11954961 on Build GRCh38
rs1028510623 Chr.1: 11955086-11955086 on Build GRCh38
rs760054077 Chr.1: 11954983-11954983 on Build GRCh38
rs953374260 Chr.1: 11954979-11954979 on Build GRCh38
rs1055842198 Chr.1: 11955176-11955176 on Build GRCh38
rs998811584 Chr.1: 11955038-11955038 on Build GRCh38
rs148869558 Chr.1: 11955096-11955096 on Build GRCh38
rs112623474 Chr.1: 11954968-11954968 on Build GRCh38
rs774767418 Chr.1: 11954907-11954907 on Build GRCh38
rs767376938 Chr.1: 11954910-11954910 on Build GRCh38
rs781376270 Chr.1: 11954867-11954867 on Build GRCh38
rs778659599 Chr.1: 11954959-11954959 on Build GRCh38
rs527751372 Chr.1: 11955152-11955152 on Build GRCh38
rs374000623 Chr.1: 11954826-11954826 on Build GRCh38
rs866816355 Chr.1: 11954934-11954934 on Build GRCh38
rs920575920 Chr.1: 11955165-11955165 on Build GRCh38
rs972314564 Chr.1: 11955159-11955159 on Build GRCh38
rs1052038445 Chr.1: 11955146-11955146 on Build GRCh38
rs577291705 Chr.1: 11954964-11954964 on Build GRCh38
rs529058654 Chr.1: 11955080-11955080 on Build GRCh38
rs371621954 Chr.1: 11954815-11954815 on Build GRCh38
rs536503346 Chr.1: 11954864-11954864 on Build GRCh38
rs751769544 Chr.1: 11955169-11955169 on Build GRCh38
rs145482271 Chr.1: 11954858-11954858 on Build GRCh38
rs906254305 Chr.1: 11955140-11955140 on Build GRCh38
rs183018672 Chr.1: 11955128-11955128 on Build GRCh38
rs199990859 Chr.1: 11954863-11954863 on Build GRCh38
rs1022596712 Chr.1: 11955210-11955210 on Build GRCh38
rs2273287 Chr.1: 11954969-11954969 on Build GRCh38
rs754057450 Chr.1: 11954927-11954928 on Build GRCh38
rs769954070 Chr.1: 11954796-11954796 on Build GRCh38
rs972027761 Chr.1: 11955061-11955061 on Build GRCh38
rs367575725 Chr.1: 11954851-11954851 on Build GRCh38
rs201291374 Chr.1: 11954853-11954853 on Build GRCh38
rs376661744 Chr.1: 11954908-11954908 on Build GRCh38
rs1014504877 Chr.1: 11954976-11954976 on Build GRCh38
rs2273288 Chr.1: 11955097-11955097 on Build GRCh38
rs775669240 Chr.1: 11954797-11954797 on Build GRCh38
rs770147701 Chr.1: 11954887-11954887 on Build GRCh38
rs201874186 Chr.1: 11954917-11954917 on Build GRCh38
rs1010808206 Chr.1: 11954975-11954975 on Build GRCh38
rs940946661 Chr.1: 11955147-11955147 on Build GRCh38
rs894558156 Chr.1: 11955178-11955178 on Build GRCh38
rs768848010 Chr.1: 11954901-11954901 on Build GRCh38
rs1011994398 Chr.1: 11954912-11954912 on Build GRCh38
rs376437784 Chr.1: 11955048-11955048 on Build GRCh38
rs116532694 Chr.1: 11955223-11955223 on Build GRCh38
rs763284492 Chr.1: 11954804-11954804 on Build GRCh38
rs998759113 Chr.1: 11955219-11955219 on Build GRCh38
rs746416126 Chr.1: 11954875-11954875 on Build GRCh38
rs534314930 Chr.1: 11954857-11954857 on Build GRCh38
rs977092079 Chr.1: 11954788-11954788 on Build GRCh38
rs557499099 Chr.1: 11954947-11954947 on Build GRCh38
rs749448103 Chr.1: 11954894-11954894 on Build GRCh38
rs903092664 Chr.1: 11955002-11955002 on Build GRCh38
rs36005914 Chr.1: 11954888-11954888 on Build GRCh38
rs1024194421 Chr.1: 11954985-11954985 on Build GRCh38
rs1030644998 Chr.1: 11954963-11954963 on Build GRCh38
rs750416079 Chr.1: 11954840-11954840 on Build GRCh38
rs899213305 Chr.1: 11954960-11954960 on Build GRCh38
rs777249474 Chr.1: 11954780-11954780 on Build GRCh38
rs767195501 Chr.1: 11954832-11954832 on Build GRCh38
rs774714468 Chr.1: 11954823-11954823 on Build GRCh38
rs753950256 Chr.1: 11955201-11955201 on Build GRCh38
rs189846231 Chr.1: 11955107-11955107 on Build GRCh38
rs775720354 Chr.1: 11954888-11954888 on Build GRCh38
rs892780668 Chr.1: 11955234-11955234 on Build GRCh38
rs774781523 Chr.1: 11955146-11955147 on Build GRCh38
rs143441347 Chr.1: 11954931-11954931 on Build GRCh38
rs985988765 Chr.1: 11954980-11954980 on Build GRCh38
rs1003259622 Chr.1: 11955141-11955141 on Build GRCh38
rs919285002 Chr.1: 11955067-11955067 on Build GRCh38
rs746335274 Chr.1: 11954783-11954783 on Build GRCh38

View Full Product Details