Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11957703-11958197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLOD1
Entrez Gene ID
5351
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 1
Gene Aliases
EDS6, LH, LH1, LLH, PLOD
UniGene
Hs.75093
Gene Chromosome Location
Chr.1: 11934667-11975542 on Build GRCh38
Transcripts
NM_000302.3 NM_001316320.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4576176 c.810C>T p.T270T Chr.1: 11957904-11957904 on Build GRCh38
7335934 c.814G>A p.E272K Chr.1: 11957914-11957914 on Build GRCh38
3704883 c.916C>T p.R306C Chr.1: 11957875-11957875 on Build GRCh38
7156623 c.930C>T p.F310F Chr.1: 11957889-11957889 on Build GRCh38
7335935 c.955G>A p.E319K Chr.1: 11957914-11957914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771746998 Chr.1: 11957833-11957833 on Build GRCh38
rs1056398577 Chr.1: 11957942-11957942 on Build GRCh38
rs766071763 Chr.1: 11957948-11957948 on Build GRCh38
rs144226170 Chr.1: 11957876-11957876 on Build GRCh38
rs756834446 Chr.1: 11957950-11957950 on Build GRCh38
rs558752942 Chr.1: 11957897-11957897 on Build GRCh38
rs755705860 Chr.1: 11957958-11957958 on Build GRCh38
rs559480062 Chr.1: 11957971-11957971 on Build GRCh38
rs779727617 Chr.1: 11957806-11957806 on Build GRCh38
rs1007349294 Chr.1: 11957987-11957987 on Build GRCh38
rs780952856 Chr.1: 11957900-11957900 on Build GRCh38
rs753603006 Chr.1: 11957810-11957810 on Build GRCh38
rs543437537 Chr.1: 11957933-11957933 on Build GRCh38
rs562232829 Chr.1: 11958003-11958003 on Build GRCh38
rs528664570 Chr.1: 11957978-11957978 on Build GRCh38
rs766349712 Chr.1: 11957930-11957930 on Build GRCh38
rs866340438 Chr.1: 11957798-11957798 on Build GRCh38
rs767557623 Chr.1: 11957799-11957799 on Build GRCh38
rs756314494 Chr.1: 11957804-11957804 on Build GRCh38
rs771071711 Chr.1: 11957979-11957979 on Build GRCh38
rs147940796 Chr.1: 11957885-11957885 on Build GRCh38
rs760869815 Chr.1: 11957926-11957926 on Build GRCh38
rs941633859 Chr.1: 11958160-11958160 on Build GRCh38
rs746658625 Chr.1: 11957837-11957837 on Build GRCh38
rs745557838 Chr.1: 11957954-11957954 on Build GRCh38
rs976931658 Chr.1: 11958099-11958099 on Build GRCh38
rs764158476 Chr.1: 11957938-11957938 on Build GRCh38
rs766692124 Chr.1: 11957873-11957873 on Build GRCh38
rs967218497 Chr.1: 11957783-11957783 on Build GRCh38
rs569632202 Chr.1: 11957836-11957836 on Build GRCh38
rs964260100 Chr.1: 11957901-11957901 on Build GRCh38
rs145447578 Chr.1: 11957905-11957905 on Build GRCh38
rs149399201 Chr.1: 11957953-11957953 on Build GRCh38
rs750459442 Chr.1: 11957863-11957863 on Build GRCh38
rs371083581 Chr.1: 11958161-11958161 on Build GRCh38
rs116133545 Chr.1: 11958163-11958163 on Build GRCh38
rs74354225 Chr.1: 11957902-11957902 on Build GRCh38
rs538763801 Chr.1: 11957864-11957864 on Build GRCh38
rs549487813 Chr.1: 11958084-11958084 on Build GRCh38
rs542453653 Chr.1: 11957994-11957994 on Build GRCh38
rs975532570 Chr.1: 11958075-11958075 on Build GRCh38
rs941436311 Chr.1: 11958155-11958155 on Build GRCh38
rs140758113 Chr.1: 11957904-11957904 on Build GRCh38
rs776949094 Chr.1: 11957932-11957932 on Build GRCh38
rs779054575 Chr.1: 11957828-11957828 on Build GRCh38
rs767610828 Chr.1: 11957861-11957861 on Build GRCh38
rs557353959 Chr.1: 11957934-11957934 on Build GRCh38
rs777431366 Chr.1: 11957894-11957894 on Build GRCh38
rs184999645 Chr.1: 11957924-11957924 on Build GRCh38
rs562008681 Chr.1: 11958115-11958115 on Build GRCh38
rs955322482 Chr.1: 11958108-11958108 on Build GRCh38
rs1047116193 Chr.1: 11957919-11957919 on Build GRCh38
rs775565324 Chr.1: 11957923-11957923 on Build GRCh38
rs752711810 Chr.1: 11957888-11957888 on Build GRCh38
rs763439166 Chr.1: 11957842-11957842 on Build GRCh38
rs908800499 Chr.1: 11957889-11957889 on Build GRCh38
rs758361490 Chr.1: 11957890-11957890 on Build GRCh38
rs983172706 Chr.1: 11958145-11958145 on Build GRCh38
rs941480174 Chr.1: 11957929-11957929 on Build GRCh38
rs532207333 Chr.1: 11958146-11958146 on Build GRCh38
rs758414759 Chr.1: 11957936-11957936 on Build GRCh38
rs373471550 Chr.1: 11957913-11957913 on Build GRCh38
rs748075257 Chr.1: 11957829-11957829 on Build GRCh38
rs776520439 Chr.1: 11957991-11957991 on Build GRCh38
rs199669594 Chr.1: 11957939-11957939 on Build GRCh38
rs768989396 Chr.1: 11957847-11957847 on Build GRCh38
rs762271097 Chr.1: 11957859-11957859 on Build GRCh38
rs1003502922 Chr.1: 11957813-11957813 on Build GRCh38
rs201244598 Chr.1: 11957960-11957960 on Build GRCh38
rs951382648 Chr.1: 11958135-11958135 on Build GRCh38
rs1022483147 Chr.1: 11957880-11957880 on Build GRCh38
rs374603327 Chr.1: 11957840-11957840 on Build GRCh38
rs760851810 Chr.1: 11958139-11958139 on Build GRCh38
rs774882355 Chr.1: 11957855-11957855 on Build GRCh38
rs368132656 Chr.1: 11957899-11957899 on Build GRCh38
rs753994916 Chr.1: 11957874-11957874 on Build GRCh38
rs531268753 Chr.1: 11958004-11958004 on Build GRCh38
rs754726612 Chr.1: 11957811-11957811 on Build GRCh38
rs1020072589 Chr.1: 11957781-11957781 on Build GRCh38
rs771755416 Chr.1: 11957736-11957736 on Build GRCh38
rs964464537 Chr.1: 11958069-11958069 on Build GRCh38
rs1023520584 Chr.1: 11957748-11957748 on Build GRCh38

View Full Product Details